Anti-DNASE1 Rabbit Polyclonal Antibody (Cy3®)
Leverandør: Bioss
BS-7651R-CY3
BOSSBS-7651R-CY3EA 10550 NOK
BOSSBS-7651R-CY3
Anti-DNASE1 Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
Deoxyribonuclease I gene is approximately 3.2 kb long with 9 exons separated by 8 introns. In the form of a bovine pancreatic enzyme preparation, it occupies an important place in the history of protein chemistry and enzymology: it was the first enzyme to be recognized as specific for DNA; it was the first DNase to be crystallized; and it was the first DNase for which a specific protein inhibitor was characterized. DNase I is a Ca2+ and Mg2+ dependant endonuclease. DNase I is synthesized in the pancreas and stored in zymogen granules. It has been used to reduce the viscosity of cysticMore Product Information