Anti-DNASE1 Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
BS-7651R-CY7
BOSSBS-7651R-CY7EA 10550 NOK
BOSSBS-7651R-CY7
Anti-DNASE1 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Deoxyribonuclease I gene is approximately 3.2 kb long with 9 exons separated by 8 introns. In the form of a bovine pancreatic enzyme preparation, it occupies an important place in the history of protein chemistry and enzymology: it was the first enzyme to be recognized as specific for DNA; it was the first DNase to be crystallized; and it was the first DNase for which a specific protein inhibitor was characterized. DNase I is a Ca2+ and Mg2+ dependant endonuclease. DNase I is synthesized in the pancreas and stored in zymogen granules. It has been used to reduce the viscosity of cysticMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol DNASE1
Environmentally Preferable
Antigen-navn Deoxyribonuclease 1
Antigen synonymer DRNI|DNL1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy7®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 1773
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at –20 °C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR