Anti-DNase 2 alpha Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-7652R-A680
BOSSBS-7652R-A680EA 10425 NOK
BOSSBS-7652R-A680
Anti-DNase 2 alpha Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Hydrolyzes DNA under acidic conditions with a preference for double-stranded DNA. Plays a major role in the degradation of nuclear DNA in cellular apoptosis during development. Necessary for proper fetal development and for definitive erythropoiesis in fetal liver, where it degrades nuclear DNA expelled from erythroid precursor cells.

Type: Primary
Antigen: DNase 2 alpha
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 25-90/360
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR