Anti-DNase 2 alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-7652R-A750
BOSSBS-7652R-A750EA 10425 NOK
BOSSBS-7652R-A750
Anti-DNase 2 alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Hydrolyzes DNA under acidic conditions with a preference for double-stranded DNA. Plays a major role in the degradation of nuclear DNA in cellular apoptosis during development. Necessary for proper fetal development and for definitive erythropoiesis in fetal liver, where it degrades nuclear DNA expelled from erythroid precursor cells.

Type: Primary
Antigen: DNase 2 alpha
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 25-90/360
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn DNase 2 alpha
Antigen synonymer Deoxyribonuclease-2-alpha|Acid DNase|DNL2|DNL|Lysosomal DNase II|DNASE2|DNase II alpha|R31240_2|Deoxyribonuclease II alpha|DNASE2A
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 750
Reaktivitet Human,Mouse
Vert Rabbit
Gen-ID 1777
Isotype IgG
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 25-90/360
Immunogen KLH conjugated synthetic peptide derived from human DNase II alpha
Skjema Liquid
Molekylvekt 38 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR