Anti-DNase gamma Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-7653R-A680
BOSSBS-7653R-A680EA 10450 NOK
BOSSBS-7653R-A680
Anti-DNase gamma Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Has DNA hydrolytic activity. Does not bind to actin. Cleaves chromatin DNA to nucleosomal units.

Type: Primary
Antigen: DNase gamma
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 151-250/305
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR