Anti-DNase gamma Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-7653R-A750
BOSSBS-7653R-A750EA 10450 NOK
BOSSBS-7653R-A750
Anti-DNase gamma Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Has DNA hydrolytic activity. Does not bind to actin. Cleaves chromatin DNA to nucleosomal units.

Type: Primary
Antigen: DNase gamma
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 151-250/305
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn DNase gamma
Antigen synonymer DNAS1L3|DNase gamma|Liver and spleen DNase|Deoxyribonuclease I-like 3|DNASE1L3|DNase I-like 3|LSD|DHP2|LS-DNase|DNase I homolog protein DHP2|SLEB16|Deoxyribonuclease gamma
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 750
Reaktivitet Rat,Mouse
Vert Rabbit
Gen-ID 1776
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 151-250/305
Immunogen KLH conjugated synthetic peptide derived from human DNase gamma
Skjema Liquid
Molekylvekt 36 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR