Anti-RACGAP1 Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-7766R
BOSSBS-7766REA 8500 NOK
BOSSBS-7766R
Anti-RACGAP1 Rabbit Polyclonal Antibody
Antistoffer
Rho GTPases control a variety of cellular processes. There are 3 subtypes of Rho GTPases in the Ras superfamily of small G proteins: RHO, RAC and CDC42. GTPase-activating proteins (GAPs) bind activated forms of Rho GTPases and stimulate GTP hydrolysis. Through this catalytic function, Rho GAPs negatively regulate Rho-mediated signals. GAPs may also serve as effector molecules and play a role in signaling downstream of Rho and other Ras-like GTPases.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: RACGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR