Anti-AICDA Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverandør: Bioss
BS-7855R-A350
BOSSBS-7855R-A350EA 10425 NOK
BOSSBS-7855R-A350
Anti-AICDA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
Single-stranded DNA-specific cytidine deaminase. Involved in somatic hypermutation, gene conversion, and class-switch recombination in B-lymphocytes. Required for several crucial steps of B-cell terminal differentiation necessary for efficient antibody responses. May also play a role in the epigenetic regulation of gene expression by participating in DNA demethylation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AICDA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR