Anti-SHOC2 Rabbit Polyclonal Antibody (Cy7®)
Leverandør: Bioss
BS-7934R-CY7
BOSSBS-7934R-CY7EA 10450 NOK
BOSSBS-7934R-CY7
Anti-SHOC2 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Regulatory subunit of protein phosphatase 1 (PP1c) that acts as a M-Ras/MRAS effector and participates in MAPK pathway activation. Upon M-Ras/MRAS activation, targets PP1c to specifically dephosphorylate the 'Ser-259' inhibitory site of RAF1 kinase and stimulate RAF1 activity at specialized signaling complexes.Involvement in disease:Defects in SHOC2 are the cause of Noonan syndrome-like with loose anagen hair (NSLAH) . NSLAH children display macrocephaly, high forehead, hypertelorism, palpebral ptosis, low-set and posteriorly rotated ears, short and webbed neck and pectus anomalies. AffectedMore Product Information