Anti-ABHD4 Rabbit Polyclonal Antibody (Cy5®)
Leverandør: Bioss
BS-7948R-CY5
BOSSBS-7948R-CY5EA 10550 NOK
BOSSBS-7948R-CY5
Anti-ABHD4 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
Lysophospholipase selective for N-acyl phosphatidylethanolamine (NAPE). Contributes to the biosynthesis of N-acyl ethanolamines, including the endocannabinoid anandamide by hydrolyzing the sn-1 and sn-2 acyl chains from N-acyl phosphatidylethanolamine (NAPE) generating glyceroN-acyl ethanolamine (GP-NAE), an intermediate for N-acyl ethanolamine biosynthesis. Hydrolyzes substrates bearing saturated, monounsaturated, polyunsaturated N-acyl chains. Shows no significant activity towards other lysophospholipids, including lysophosphatidylcholine, lysophosphatidylethanolamine andMore Product Information