Anti-AOC1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Leverandør: Bioss
BS-7949R-A488
BOSSBS-7949R-A488EA 10450 NOK
BOSSBS-7949R-A488
Anti-AOC1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Catalyzes the degradation of compounds such as putrescine, histamine, spermine, and spermidine, substances involved in allergic and immune responses, cell proliferation, tissue differentiation, tumor formation, and possibly apoptosis. Placental DAO is thought to play a role in the regulation of the female reproductive function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AOC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR