Anti-DAO1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-7949R-A680
BOSSBS-7949R-A680EA 10450 NOK
BOSSBS-7949R-A680
Anti-DAO1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Catalyzes the degradation of compounds such as putrescine, histamine, spermine, and spermidine, substances involved in allergic and immune responses, cell proliferation, tissue differentiation, tumor formation, and possibly apoptosis. Placental DAO is thought to play a role in the regulation of the female reproductive function.

Type: Primary
Antigen: DAO1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 601-689/751
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR