Anti-AKR1A1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Leverandør: Bioss
BS-7953R-A680
BOSSBS-7953R-A680EA 10275 NOK
BOSSBS-7953R-A680
Anti-AKR1A1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Catalyzes the NADPH-dependent reduction of a variety of aromatic and aliphatic aldehydes to their corresponding alcohols. Catalyzes the reduction of mevaldate to mevalonic acid and of glyceraldehyde to glycerol. Has broad substrate specificity. In vitro substrates include succinic semialdehyde, 4-nitrobenzaldehyde, 1,2-naphthoquinone, methylglyoxal, and D-glucuronic acid. Plays a role in the activation of procarcinogens, such as polycyclic aromatic hydrocarbon trans-dihydrodiols, and in the metabolism of various xenobiotics and drugs, including the anthracyclines doxorubicin (DOX) andMore Product Information