Anti-HIAT1 Rabbit Polyclonal Antibody (Cy3®)
Leverandør: Bioss
BS-8042R-CY3
BOSSBS-8042R-CY3EA 10550 NOK
BOSSBS-8042R-CY3
Anti-HIAT1 Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
The Major facilitator superfamily consists of presumed carbohydrate transporters with 10-12 membrane-spanning domains. Belonging to the facilitator superfamily, HIAT1 is a 490 amino acid multi-pass membrane protein that may function as a sugar transporter and is expressed in adult and embryonic brain. The HIAT1 gene was first observed while analyzing for active genes in neonatal mouse hippocampus. The gene encoding HIAT1 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes onMore Product Information