Anti-HIAT1 Rabbit Polyclonal Antibody (Cy5®)

Leverandør: Bioss
BS-8042R-CY5
BOSSBS-8042R-CY5EA 10550 NOK
BOSSBS-8042R-CY5
Anti-HIAT1 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
The Major facilitator superfamily consists of presumed carbohydrate transporters with 10-12 membrane-spanning domains. Belonging to the facilitator superfamily, HIAT1 is a 490 amino acid multi-pass membrane protein that may function as a sugar transporter and is expressed in adult and embryonic brain. The HIAT1 gene was first observed while analyzing for active genes in neonatal mouse hippocampus. The gene encoding HIAT1 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes onMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol HIAT1
Environmentally Preferable
Antigen-navn Hippocampus Abundant Transcript 1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 64645
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at –20 °C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR