Anti-HIAT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverandør: Bioss
BS-8042R-HRP
BOSSBS-8042R-HRPEA 10550 NOK
BOSSBS-8042R-HRP
Anti-HIAT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
The Major facilitator superfamily consists of presumed carbohydrate transporters with 10-12 membrane-spanning domains. Belonging to the facilitator superfamily, HIAT1 is a 490 amino acid multi-pass membrane protein that may function as a sugar transporter and is expressed in adult and embryonic brain. The HIAT1 gene was first observed while analyzing for active genes in neonatal mouse hippocampus. The gene encoding HIAT1 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes onMore Product Information