Anti-FAM50A Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-8152R
BOSSBS-8152REA 8500 NOK
BOSSBS-8152R
Anti-FAM50A Rabbit Polyclonal Antibody
Antistoffer
FAM50A, also known as DXS9928E, HXC26, XAP5 or 9F, is a 339 amino acid nuclear protein that belongs to the FAM50 family. Expressed ubiquitously with highest expression in fetal kidney, liver and brain, as well as adult heart, spleen, skeletal muscle, prostate and small intestine, FAM50A is thought to function as a transcription factor that may bind to DNA. FAM50A contains an SV40 large T antigen nuclear localization signal and a polymorphic CCG repeat region in its 5’-UTR. Defects in the gene encoding FAM50A may be associated with acute lymphoblastic leukemia, suggesting a possible roleMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol FAM50A
Antigen-navn Family with sequence similarity 50, member A
Antigen synonymer HXC-26|XAP5|HXC26|9F|DXS9928E
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 9130
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A.
Konsentrasjon 1 ug/ul
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR