Anti-GNAT1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverandør: Bioss
BS-8160R-A350
BOSSBS-8160R-A350EA 10450 NOK
BOSSBS-8160R-A350
Anti-GNAT1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
GTP-binding proteins (G-proteins)are a family of heterotrimeric proteins that play a critical role in signal transduction by coupling cell surface, 7-transmembrane domain receptors to intracellular signaling pathways including second messenger generation (such as cyclic AMP, calcium and diacylglycerol), protein phosphorylation, ion channel activation, gene induction, cell growth and differentiation. Receptor activation catalyzes the exchange of GTP for GDP bound to the inactive G protein alpha subunit resulting in a conformational change and dissociation of the complex. The G protein alphaMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol GNAT1
Antigen-navn Guanine Nucleotide Binding Protein (G Protein) Alpha Transducing Activity Polypeptide 1
Environmentally Preferable
Antigen synonymer CSNB1G|CSNBAD3|GBT1|GNATR
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 350
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 2779
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at –20 °C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR