Anti-GNAT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Leverandør: Bioss
BS-8160R-FITC
BOSSBS-8160R-FITCEA 10550 NOK
BOSSBS-8160R-FITC
Anti-GNAT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
GTP-binding proteins (G-proteins)are a family of heterotrimeric proteins that play a critical role in signal transduction by coupling cell surface, 7-transmembrane domain receptors to intracellular signaling pathways including second messenger generation (such as cyclic AMP, calcium and diacylglycerol), protein phosphorylation, ion channel activation, gene induction, cell growth and differentiation. Receptor activation catalyzes the exchange of GTP for GDP bound to the inactive G protein alpha subunit resulting in a conformational change and dissociation of the complex. The G protein alphaMore Product Information