Anti-PHF8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Leverandør: Bioss
BS-8176R-A488
BOSSBS-8176R-A488EA 10450 NOK
BOSSBS-8176R-A488
Anti-PHF8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
The protein encoded by this gene is a histone lysine demethylase that preferentially acts on histones in the monomethyl or dimethyl states. The encoded protein requires Fe(2+) ion, 2-oxoglutarate, and oxygen for its catalytic activity. Defects in this gene are a cause of mental retardation syndromic X-linked Siderius type (MRXSSD). Four transcript variants encoding different isoforms have been found for this gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PHF8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol PHF8
Antigen-navn PHD Finger Protein 8
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 488
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 23133
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at –20 °C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR