Anti-PHF8 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverandør: Bioss
BS-8176R-A555
BOSSBS-8176R-A555EA 10450 NOK
BOSSBS-8176R-A555
Anti-PHF8 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
The protein encoded by this gene is a histone lysine demethylase that preferentially acts on histones in the monomethyl or dimethyl states. The encoded protein requires Fe(2+) ion, 2-oxoglutarate, and oxygen for its catalytic activity. Defects in this gene are a cause of mental retardation syndromic X-linked Siderius type (MRXSSD). Four transcript variants encoding different isoforms have been found for this gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PHF8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR