Anti-PHF8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-8176R-A680
BOSSBS-8176R-A680EA 10450 NOK
BOSSBS-8176R-A680
Anti-PHF8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
The protein encoded by this gene is a histone lysine demethylase that preferentially acts on histones in the monomethyl or dimethyl states. The encoded protein requires Fe(2+) ion, 2-oxoglutarate, and oxygen for its catalytic activity. Defects in this gene are a cause of mental retardation syndromic X-linked Siderius type (MRXSSD). Four transcript variants encoding different isoforms have been found for this gene.

Type: Primary
Antigen: PHF8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 501-600/1060
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigen-navn PHF8
Antigen synonymer PHF8|Histone lysine demethylase PHF8|ZNF422|PHD finger protein 8|PHF8_HUMAN
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 680
Reaktivitet Human
Vert Rabbit
Gen-ID 23133
Isotype IgG
Flowcytometri Yes
Modifikasjon Unmodified
Epitop 501-600/1060
Immunogen KLH conjugated synthetic peptide derived from human PHF8
Skjema Liquid
Molekylvekt 118 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR