Anti-PHF8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-8176R-A750
BOSSBS-8176R-A750EA 10450 NOK
BOSSBS-8176R-A750
Anti-PHF8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
The protein encoded by this gene is a histone lysine demethylase that preferentially acts on histones in the monomethyl or dimethyl states. The encoded protein requires Fe(2+) ion, 2-oxoglutarate, and oxygen for its catalytic activity. Defects in this gene are a cause of mental retardation syndromic X-linked Siderius type (MRXSSD). Four transcript variants encoding different isoforms have been found for this gene.

Type: Primary
Antigen: PHF8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 501-600/1060
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR