Anti-FAM161A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Leverandør: Bioss
BS-8216R-A680
BOSSBS-8216R-A680EA 10275 NOK
BOSSBS-8216R-A680
Anti-FAM161A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Isoform 1 and isoform 3 are widely expressed with highest levels in retina and testis, with isoform 1 being the mot abundant in all tissues tested.Involvement in disease:Defects in FAM161A are the cause of retinitis pigmentosa type 28 (RP28) . A retinal dystrophy belonging to the group of pigmentary retinopathies. Retinitis pigmentosa is characterized by retinal pigment deposits visible on fundus examination and primary loss of rod photoreceptor cells followed by secondary loss of cone photoreceptors. Patients typically have night vision blindness and loss of midperipheral visual field. AsMore Product Information