Anti-DHPS Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Leverandør: Bioss
BS-8259R-A488
BOSSBS-8259R-A488EA 10450 NOK
BOSSBS-8259R-A488
Anti-DHPS Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Catalyzes the NAD-dependent oxidative cleavage of spermidine and the subsequent transfer of the butylamine moiety of spermidine to the epsilon-amino group of a specific lysine residue of the eIF-5A precursor protein to form the intermediate deoxyhypusine residue.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DHPS
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR