Anti-DHPS Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverandør: Bioss
BS-8259R-A555
BOSSBS-8259R-A555EA 10450 NOK
BOSSBS-8259R-A555
Anti-DHPS Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
Catalyzes the NAD-dependent oxidative cleavage of spermidine and the subsequent transfer of the butylamine moiety of spermidine to the epsilon-amino group of a specific lysine residue of the eIF-5A precursor protein to form the intermediate deoxyhypusine residue.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DHPS
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol DHPS
Environmentally Preferable
Antigen-navn deoxyhypusine synthase
Antigen synonymer DS|MIG13|DHS
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 555
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 1725
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at –20 °C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR