Anti-DHPS Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-8259R-A750
BOSSBS-8259R-A750EA 10450 NOK
BOSSBS-8259R-A750
Anti-DHPS Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Catalyzes the NAD-dependent oxidative cleavage of spermidine and the subsequent transfer of the butylamine moiety of spermidine to the epsilon-amino group of a specific lysine residue of the eIF-5A precursor protein to form the intermediate deoxyhypusine residue.

Type: Primary
Antigen: DHPS
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 111-200/369
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR