Anti-SERPINB13 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverandør: Bioss
BS-8324R-A555
BOSSBS-8324R-A555EA 10450 NOK
BOSSBS-8324R-A555
Anti-SERPINB13 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
Headpin is a skin-specific, UV-repressible serine proteinase inhibitor (serpin) belonging to the ovalbumin serpin family. Headpin is abundant in the human keratinocyte cell line HaCaT, and in lesional keratinocytes from psoriatic skin. Headpin downregulation occurs in squamous cell carcinoma of the oral cavity and in squamous cell carcinoma cell lines of the head and neck.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SERPINB13
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR