Anti-SERPINB13 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-8324R-A680
BOSSBS-8324R-A680EA 10450 NOK
BOSSBS-8324R-A680
Anti-SERPINB13 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Headpin is a skin-specific, UV-repressible serine proteinase inhibitor (serpin) belonging to the ovalbumin serpin family. Headpin is abundant in the human keratinocyte cell line HaCaT, and in lesional keratinocytes from psoriatic skin. Headpin downregulation occurs in squamous cell carcinoma of the oral cavity and in squamous cell carcinoma cell lines of the head and neck.

Type: Primary
Antigen: SERPINB13
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/391
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn SERPINB13
Antigen synonymer PI13|serpinB13|Hurpin|Protease inhibitor 13|Peptidase inhibitor 13|HaCaT UV-repressible serpin|Headpin|Proteinase inhibitor 13|Serpin B13|Serpin peptidase inhibitor clade B ovalbumin member 13|SPB13_HUMAN|HUR7|HSHUR7SEQ|Serine or cysteine proteinase inhibitor clade B ovalbumin member 13|SERPINB13|PI-13|UV B repressed sequence HUR 7
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 680
Reaktivitet
Vert Rabbit
Gen-ID 5275
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 101-200/391
Immunogen KLH conjugated synthetic peptide derived from human Headpin/SERPINB13
Skjema Liquid
Molekylvekt 44 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR