Anti-SERPINB13 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-8324R-A750
BOSSBS-8324R-A750EA 10450 NOK
BOSSBS-8324R-A750
Anti-SERPINB13 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Headpin is a skin-specific, UV-repressible serine proteinase inhibitor (serpin) belonging to the ovalbumin serpin family. Headpin is abundant in the human keratinocyte cell line HaCaT, and in lesional keratinocytes from psoriatic skin. Headpin downregulation occurs in squamous cell carcinoma of the oral cavity and in squamous cell carcinoma cell lines of the head and neck.

Type: Primary
Antigen: SERPINB13
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/391
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR