Anti-STIM1 Rabbit Polyclonal Antibody (Cy3®)

Leverandør: Bioss
BS-8526R-CY3
BOSSBS-8526R-CY3EA 10550 NOK
BOSSBS-8526R-CY3
Anti-STIM1 Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
Plays a role in mediating store-operated Ca(2+) entry (SOCE), a Ca(2+) influx following depletion of intracellular Ca(2+) stores. Acts as Ca(2+) sensor in the endoplasmic reticulum via its EF-hand domain. Upon Ca(2+) depletion, translocates from the endoplasmic reticulum to the plasma membrane where it activates the Ca(2+) release-activated Ca(2+) (CRAC) channel subunit, TMEM142A/ORAI1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STIM1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR