Anti-AKAP2 Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
BS-9028R-CY7
BOSSBS-9028R-CY7EA 10550 NOK
BOSSBS-9028R-CY7
Anti-AKAP2 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Binds to regulatory subunit (RII) of protein kinase A. May be involved in establishing polarity in signaling systems or in integrating PKA-RII isoforms with downstream effectors to capture, amplify and focus diffuse, trans-cellular signals carried by cAMP (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AKAP2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol AKAP2
Environmentally Preferable
Antigen-navn AKAP2
Antigen synonymer AKAP-2|PRKA2|AKAPKL
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy7®
Reaktivitet Human,Mouse
Vert Rabbit
Gen-ID 11217
Isotype IgG
ImmunoKjemi Yes
Immunogen 165-215/859
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at –20 °C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR