Anti-TRPM7 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Leverandør: Bioss
BS-9044R-A555
BOSSBS-9044R-A555EA 10450 NOK
BOSSBS-9044R-A555
Anti-TRPM7 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
Essential ion channel and serine/threonine-protein kinase. Divalent cation channel permeable to calcium and magnesium. Has a central role in magnesium ion homeostasis and in the regulation of anoxic neuronal cell death. Involved in TNF-induced necroptosis downstream of MLKL by mediating calcium influx. The kinase activity is essential for the channel function. May be involved in a fundamental process that adjusts plasma membrane divalent cation fluxes according to the metabolic state of the cell. Phosphorylates annexin A1 (ANXA1).
Recommended Dilutions: IF(IHC-P): 1:50-200