Anti-TRPM7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-9044R-A750
BOSSBS-9044R-A750EA 10450 NOK
BOSSBS-9044R-A750
Anti-TRPM7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Essential ion channel and serine/threonine-protein kinase. Divalent cation channel permeable to calcium and magnesium. Has a central role in magnesium ion homeostasis and in the regulation of anoxic neuronal cell death. Involved in TNF-induced necroptosis downstream of MLKL by mediating calcium influx. The kinase activity is essential for the channel function. May be involved in a fundamental process that adjusts plasma membrane divalent cation fluxes according to the metabolic state of the cell. Phosphorylates annexin A1 (ANXA1).

Type: Primary
Antigen: TRPM7
Clonality: Polyclonal
Clone:
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR