Anti-TRPM7 Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
BS-9044R-CY7
BOSSBS-9044R-CY7EA 10550 NOK
BOSSBS-9044R-CY7
Anti-TRPM7 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Essential ion channel and serine/threonine-protein kinase. Divalent cation channel permeable to calcium and magnesium. Has a central role in magnesium ion homeostasis and in the regulation of anoxic neuronal cell death. Involved in TNF-induced necroptosis downstream of MLKL by mediating calcium influx. The kinase activity is essential for the channel function. May be involved in a fundamental process that adjusts plasma membrane divalent cation fluxes according to the metabolic state of the cell. Phosphorylates annexin A1 (ANXA1).

Recommended Dilutions: IF(IHC-P): 1:50-200

Bestill nå

SPESIFIKASJONER

Antigensymbol TRPM7
Antigen-navn Transient Receptor Potential Cation Channel Subfamily M member 7
Environmentally Preferable
Antigen synonymer TRPPLIK|CHAK1|LTrpC-7|2310022G15Rik|Ltrpc7|Ltpr7|CHAK|4833414K03Rik|5033407O22Rik
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy7®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 54822
Isotype IgG
ImmunoKjemi Yes
Immunogen 800-850/1865
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at –20 °C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR