Anti-ZNRF2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-9140R-A750
BOSSBS-9140R-A750EA 10450 NOK
BOSSBS-9140R-A750
Anti-ZNRF2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). ZNRF2 (zinc and ring finger 2), also known as RNF202, is a 242 amino acid peripheral membrane protein that contains one RING-type zinc finger and localises to the lysosome, as well as the endosome and the cell junction. Expressed at high levels in brain tissue,More Product Information
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn ZNRF2
Antigen synonymer Protein Ells2|Zinc/RING finger protein 2|Znrf2|E3 ubiquitin-protein ligase ZNRF2|ZNRF2_HUMAN|RING finger protein 202
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 750
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 223082
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 151-242/242
Immunogen KLH conjugated synthetic peptide derived from human ZNRF2
Skjema Liquid
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR