Anti-ZNRF2 Rabbit Polyclonal Antibody (Cy5®)

Leverandør: Bioss
BS-9140R-CY5
BOSSBS-9140R-CY5EA 10550 NOK
BOSSBS-9140R-CY5
Anti-ZNRF2 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). ZNRF2 (zinc and ring finger 2), also known as RNF202, is a 242 amino acid peripheral membrane protein that contains one RING-type zinc finger and localizes to the lysosome, as well as the endosome and the cell junction. Expressed at high levels in brain tissue,More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol ZNRF2
Antigen-navn Zinc And Ring Finger 2
Environmentally Preferable
Antigen synonymer RNF202|Protein Ells2|Zinc/RING finger protein 2|Znrf2|ZNRF2_HUMAN.|E3 ubiquitin-protein ligase ZNRF2|RING finger protein 202
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 223082
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at –20 °C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR