Anti-RNF142 Rabbit Polyclonal Antibody (Cy5.5®)

Leverandør: Bioss
BS-9156R-CY5.5
BOSSBS-9156R-CY5.5EA 10550 NOK
BOSSBS-9156R-CY5.5
Anti-RNF142 Rabbit Polyclonal Antibody (Cy5.5®)
Antistoffer
SH3MD2 acts as a scaffold protein, contributes to Rac-induced signal transduction such as JNKs (MAPK8 and MAPK9) activation and induces apoptosis. Within a signaling complex, it probably recruits protein kinases such as MAP3K10 or MAP3K11 which are in turn activated leading to the sequential activation of MAP2K4, MAP2K7 and JNKs (MAPK8 and MAPK9). SH3MD2 may be involved in targeting of HIV-1 GAG and GAG-POL polyproteins to the plasma membrane. This gene encodes a protein containing an N-terminus RING-finger, four SH3 domains, and a region implicated in binding of the Rho GTPase Rac. Via theMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol RNF142
Environmentally Preferable
Antigen-navn Ring Finger Protein 142
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5.5®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 57630
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at –20 °C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR