Anti-RNF142 Rabbit Polyclonal Antibody (Cy7®)
Leverandør: Bioss
BS-9156R-CY7
BOSSBS-9156R-CY7EA 10550 NOK
BOSSBS-9156R-CY7
Anti-RNF142 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
SH3MD2 acts as a scaffold protein, contributes to Rac-induced signal transduction such as JNKs (MAPK8 and MAPK9) activation and induces apoptosis. Within a signaling complex, it probably recruits protein kinases such as MAP3K10 or MAP3K11 which are in turn activated leading to the sequential activation of MAP2K4, MAP2K7 and JNKs (MAPK8 and MAPK9). SH3MD2 may be involved in targeting of HIV-1 GAG and GAG-POL polyproteins to the plasma membrane. This gene encodes a protein containing an N-terminus RING-finger, four SH3 domains, and a region implicated in binding of the Rho GTPase Rac. Via theMore Product Information