Anti-5 MethylCytosine Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-9450R-A680
BOSSBS-9450R-A680EA 10450 NOK
BOSSBS-9450R-A680
Anti-5 MethylCytosine Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Cytidine is a nucleoside formed by a cytosine attached to a ribose ring via a beta-N1-glycosidic bond. DNA is methylated on cytidines by DNA methylases (DNMTs)to generate 5-methylcytidine (5-mC), a potent epigenetics marker and regulator of gene expression. The reverse reaction (cytidine demethylation) starts with its oxidation to hydroxymethyl- (5-hmC), formyl- (5-fC), and carboxy- (5-caC) cytidine. Several enzymes, including the Tet family of proteins have been implicated in cytidine demethylation.

Type: Primary
Antigen: 5 MethylCytosine
Clonality: Polyclonal
Clone:
Conjugation: ALEXAMore Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR