Anti-F12 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverandør: Bioss
BS-9503R-A350
BOSSBS-9503R-A350EA 10450 NOK
BOSSBS-9503R-A350
Anti-F12 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
This gene encodes coagulation factor XII which circulates in blood as a zymogen. This single chain zymogen is converted to a two-chain serine protease with an heavy chain (alpha-factor XIIa) and a light chain. The heavy chain contains two fibronectin-type domains, two epidermal growth factor (EGF)-like domains, a kringle domain and a proline-rich domain, whereas the light chain contains only a catalytic domain. On activation, further cleavages takes place in the heavy chain, resulting in the production of beta-factor XIIa light chain and the alpha-factor XIIa light chain becomes beta-factorMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol F12
Antigen-navn Coagulation Factor 12
Environmentally Preferable
Antigen synonymer HAF|Coagulation Factor XII|HAE3|Hageman Factor|HAEX
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 350
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 2161
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at –20 °C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR