Anti-Factor 12 heavy chain Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-9503R-A750
BOSSBS-9503R-A750EA 10450 NOK
BOSSBS-9503R-A750
Anti-Factor 12 heavy chain Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
This gene encodes coagulation factor XII which circulates in blood as a zymogen. This single chain zymogen is converted to a two-chain serine protease with an heavy chain (alpha-factor XIIa) and a light chain. The heavy chain contains two fibronectin-type domains, two epidermal growth factor (EGF)-like domains, a kringle domain and a proline-rich domain, whereas the light chain contains only a catalytic domain. On activation, further cleavages takes place in the heavy chain, resulting in the production of beta-factor XIIa light chain and the alpha-factor XIIa light chain becomes beta-factorMore Product Information
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn Factor 12 heavy chain
Antigen synonymer Coagulation factor XII|HAF|Factor XII deficiency|Factor XII|HAE3|F12 deficiency|HAF deficiency|Factor XII heavy chain|FA12_HUMAN|Coagulation factor XIIa heavy chain|Hageman factor|F12|HAEX
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 750
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 2161
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 31-130/615
Immunogen KLH conjugated synthetic peptide derived from human Coagulation factor XIIa heavy chain
Skjema Liquid
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR