Anti-F12 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverandør: Bioss
BS-9503R-HRP
BOSSBS-9503R-HRPEA 10550 NOK
BOSSBS-9503R-HRP
Anti-F12 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
This gene encodes coagulation factor XII which circulates in blood as a zymogen. This single chain zymogen is converted to a two-chain serine protease with an heavy chain (alpha-factor XIIa) and a light chain. The heavy chain contains two fibronectin-type domains, two epidermal growth factor (EGF)-like domains, a kringle domain and a proline-rich domain, whereas the light chain contains only a catalytic domain. On activation, further cleavages takes place in the heavy chain, resulting in the production of beta-factor XIIa light chain and the alpha-factor XIIa light chain becomes beta-factorMore Product Information