Anti-CLNS1A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Leverandør: Bioss
BS-9851R-A750
BOSSBS-9851R-A750EA 10450 NOK
BOSSBS-9851R-A750
Anti-CLNS1A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
The formation of the spliceosome includes the assembly of Sm proteins in an ordered manner onto snRNAs. This process is mediated by the survival of motor neuron (SMN) protein, and is enhanced by modification of specific arginine residues in the Sm proteins to symmetrical dimethylarginines (sDMAs). sDMA modification of Sm proteins is catalysed by the methylosome, a complex comprised of the type II methyltransferase PRMT5 (also designated Jak-binding protein 1, JBP1), pICln, and two novel factors. PRMT5 binds the Sm proteins via their arginine- and glycine-rich (RG) domains, while pICln bindsMore Product Information