2,4-D Mouse Monoclonal Antibody [clone: 24D1] (ALEXA FLUOR® 680)

Leverandør: Bioss
BSM-0861M-A680
BOSSBSM-0861M-A680EA 12650 NOK
BOSSBSM-0861M-A680
2,4-D Mouse Monoclonal Antibody [clone: 24D1] (ALEXA FLUOR® 680)
Antistoffer
2,4-Dichlorophenoxyacetic acid (2,4-D) is a common systemic herbicide used in the control of broadleaf weeds. It is the most widely used herbicide in the world, and the third most commonly used in North America. 2,4-D is also an important synthetic auxin, often used in laboratories for plant research and as a supplement in plant cell culture media such as MS medium. 2,4-D is a synthetic auxin, which is a class of plant growth regulators. It is absorbed through the leaves and is translocated to the meristems of the plant. Uncontrolled, unsustainable growth ensues causing stem curl-over, leafMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol 2,4-D
Antigen-navn 2,4-Dichlorophenoxyacetic acid
Environmentally Preferable
Antigen synonymer 2,4-Dichlorophenoxyacetic acid
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Alexa Fluor® 680
Klon 24D1
Reaktivitet
Vert Mouse
Isotype IgG
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Immunogen KLH conjugated to 2,4-D
Skjema Liquid
Rensing Purified by Protein G
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR