AFP Mouse Monoclonal Antibody [clone: A2] (ALEXA FLUOR® 750)
Leverandør: Bioss
BSM-1621M-A750
BOSSBSM-1621M-A750EA 13125 NOK
BOSSBSM-1621M-A750
AFP Mouse Monoclonal Antibody [clone: A2] (ALEXA FLUOR® 750)
Antistoffer
Alpha-fetoprotein (AFP) is a major plasma protein produced by the yolk sac and the liver during fetal life. Alpha-fetoprotein expression in adults is often associated with hepatoma or teratoma. However, hereditary persistance of alpha-fetoprotein may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and the alpha-fetoprotein and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. Alpha-fetoprotein is found in monomeric as well as dimeric and trimeric forms, and binds copper,More Product Information