AFP Mouse Monoclonal Antibody [clone: A4] (ALEXA FLUOR® 680)

Leverandør: Bioss
BSM-1622M-A680
BOSSBSM-1622M-A680EA 13125 NOK
BOSSBSM-1622M-A680
AFP Mouse Monoclonal Antibody [clone: A4] (ALEXA FLUOR® 680)
Antistoffer
Binds copper, nickel, and fatty acids as well as, and bilirubin less well than, serum albumin. Only a small percentage (less than 2%) of the human AFP shows estrogen-binding properties.

Type: Primary
Antigen: Alpha-fetoprotein
Clonality: Monoclonal
Clone: A4
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR