Anti-alpha Estradiol Mouse Monoclonal Antibody [clone: 2G2] (ALEXA FLUOR® 750)

Leverandør: Bioss
BSM-4533M-A750
BOSSBSM-4533M-A750EA 12650 NOK
BOSSBSM-4533M-A750
Anti-alpha Estradiol Mouse Monoclonal Antibody [clone: 2G2] (ALEXA FLUOR® 750)
Antistoffer
Estradiol (known as alpha Estradiol or 17 alpha Estradiol) is a biologically active estrogen in human breast cancer cells in tissue culture.17-estradiol and its selective receptor, ER-X, are not part of a classical hormone/receptor endocrine system but of a system with important autocrine/paracrine functions in the developing and adult brain. 17-Estradiol may have enormous implications for hormone replacement strategies at the menopause and in the treatment of such neurodegenerative disorders as Alzheimer?s disease and ischemic stroke.

Type: Primary
Antigen: alpha Estradiol
Clonality:More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR