Sterican® Mix Blunt Fill Needles
Leverandør: B. Braun
4038088B
BRAU4038088BEA 215 NOK
BRAU4038088B
Sterican® Mix Blunt Fill Needles
Sprøytenåler
Ideal for drug admixture.
  • 40° and 90° blunt fill needles offer clinicians flexibility
  • Blunt and semi blunt needles minimises the risk of needlestick injuries
  • Thin walled cannula allows for faste medication draw up
Pink coloured hub and protective cap for blunt fill needles.
Red coloured hub and protective cap for semi blunt needles.