Infusion filter, Sterifix®
Leverandør: B. Braun
4099303
BRAU4099303EA 10850 NOK
BRAU4099303
Infusion filter, Sterifix®
Infusjoner
Sterifix infusion filter is specially designed for bacteria and particle retention.
  • Reliable air venting regardless of position
  • High throughput 0,2 μm Supor® membrane
  • Pressure resistant to 3,1 bar Luer lock fitting
  • Low absorption rate of proteins